Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Prx
Entrez Gene ID
19153
Gene Name
periaxin
Gene Aliases
L-Periaxin
Chromosome Location
Chr.7: 27499305 - 27520041 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_198048.2 NP_932165.2 6 - 7 662 97
GenBank mRNA AJ222968.1 - 6 - 7 661 97
AK173220.1 - 2 - 3 459 97
BC068135.1 - 2 - 3 475 97

View Full Product Details