Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Slc5a4a
Entrez Gene ID
64452
Gene Name
solute carrier family 5, member 4a
Gene Aliases
SGLT3-a
Chromosome Location
Chr.10: 76147451 - 76189265 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_133184.2 NP_573447.2 10 - 11 1209 63

View Full Product Details