Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Prmt2
Entrez Gene ID
15468
Gene Name
protein arginine N-methyltransferase 2
Gene Aliases
AI504737, Hrmt1l1
Chromosome Location
Chr.10: 76207226 - 76237865 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AF169620.1 - 7 - 8 947 63
AK077456.1 - 7 - 8 1004 63
AK140200.1 - 8 - 9 1092 63
AK144412.1 - 7 - 8 1002 63
AK145829.1 - 7 - 8 1004 63
AK146754.1 - 8 - 9 1065 63
AK146966.1 - 7 - 8 1011 63
AK159186.1 - 7 - 8 1010 63
AK188145.1 - 1 - 2 95 63
BC088997.1 - 7 - 8 961 63
BC122563.1 - 6 - 7 935 63
BC125275.1 - 6 - 7 935 63

View Full Product Details