Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Mmp11
Entrez Gene ID
17385
Gene Name
matrix metallopeptidase 11
Gene Aliases
MMP-11, SL-3, ST3, Stmy3
Chromosome Location
Chr.10: 75923221 - 75932520 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK165778.1 - 8 - 9 1521 65

View Full Product Details