Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rora
Entrez Gene ID
19883
Gene Name
RAR-related orphan receptor alpha
Gene Aliases
9530021D13Rik, Nr1f1, ROR1, ROR2, ROR3, nmf267, sg, staggerer, tmgc26
Chromosome Location
Chr.9: 68653402 - 69388246 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001289916.1 NP_001276845.1 9 - 10 1411 68
NM_013646.2 NP_038674.1 10 - 11 1420 68
GenBank mRNA AK043990.1 - 6 - 7 780 68
AK149294.1 - 9 - 10 1301 68
AK155827.1 - 9 - 10 1362 68
BC003757.1 - 9 - 10 1360 68
S82720.1 - 9 - 10 1334 68
U53228.1 - 10 - 11 1404 68
Y08640.1 - 9 - 10 1566 68
Z82994.1 - 9 - 10 1399 68

View Full Product Details