Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rora
Entrez Gene ID
19883
Gene Name
RAR-related orphan receptor alpha
Gene Aliases
9530021D13Rik, Nr1f1, ROR1, ROR2, ROR3, nmf267, sg, staggerer, tmgc26
Chromosome Location
Chr.9: 68653402 - 69388246 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001289916.1 NP_001276845.1 6 - 7 1089 100
NM_013646.2 NP_038674.1 7 - 8 1098 100
GenBank mRNA AK043990.1 - 3 - 4 458 100
AK149294.1 - 6 - 7 979 100
AK155827.1 - 6 - 7 1040 100
BC003757.1 - 6 - 7 1038 100
S82720.1 - 6 - 7 1012 100
U53228.1 - 7 - 8 1082 100
Y08640.1 - 6 - 7 100
Z82994.1 - 6 - 7 1077 100

View Full Product Details