Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ccdc82
Entrez Gene ID
66396
Gene Name
coiled-coil domain containing 82
Gene Aliases
2310043N13Rik
Chromosome Location
Chr.9: 13246979 - 13292353 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_025534.2 NP_079810.2 4 - 5 1061 125
GenBank mRNA BC040405.1 - 2 - 3 222 125
BC057326.1 - 4 - 5 1061 125
BC098496.1 - 3 - 4 988 125

View Full Product Details