Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Thy1
Entrez Gene ID
21838
Gene Name
thymus cell antigen 1, theta
Gene Aliases
CD90, T25, Thy-1, Thy-1.2, Thy1.1, Thy1.2
Chromosome Location
Chr.9: 44043384 - 44048579 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_009382.3 NP_033408.1 1 - 2 93 61
GenBank mRNA CX123560.1 - 1 - 2 93 61

View Full Product Details