Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Lctl
Entrez Gene ID
235435
Gene Name
lactase-like
Gene Aliases
E130104I05Rik, KLPH
Chromosome Location
Chr.9: 64117123 - 64138118 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA BC030631.1 - 9 - 10 1189 76

View Full Product Details