The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001102468.1 | NP_001095938.1 | 3 - 4 | 282 | 67 |
NM_138304.2 | NP_612177.1 | 4 - 5 | 423 | 67 | |
GenBank mRNA | AK008039.1 | - | 4 - 5 | 409 | 67 |
AY061807.1 | - | 4 - 5 | 423 | 67 | |
BC013541.1 | - | 4 - 5 | 401 | 67 | |
BC023475.1 | - | 4 - 5 | 388 | 67 | |
CO039047.1 | - | 1 - 2 | 67 |