Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Olfr77
Entrez Gene ID
258336
Gene Name
olfactory receptor 77
Gene Aliases
18A, MOR143-1
Chromosome Location
Chr.9: 19917229 - 19922510 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_146339.3 NP_666451.1 1 - 2 213 155
GenBank mRNA BC046590.1 - 1 - 2 213 155

View Full Product Details