Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Gramd2
Entrez Gene ID
546134
Gene Name
GRAM domain containing 2
Gene Aliases
BC064463
Chromosome Location
Chr.9: 59679462 - 59718871 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001033498.1 NP_001028670.1 5 - 6 666 59
GenBank mRNA AK040545.1 - 6 - 7 555 59
AK041631.1 - 5 - 6 666 59
BC138948.1 - 5 - 6 483 59
BC138949.1 - 5 - 6 483 59

View Full Product Details