The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_027823.1 | NP_082099.1 | 21 - 22 | 2592 | 61 |
GenBank mRNA | BC137891.1 | - | 21 - 22 | 2592 | 61 |
BC137892.1 | - | 21 - 22 | 2592 | 61 |