Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Pvrl1
Entrez Gene ID
58235
Gene Name
poliovirus receptor-related 1
Gene Aliases
AI835281, AW549174, Cd111, HIgR, HveC, PRR, PRR1, nectin-1
Chromosome Location
Chr.9: 43744576 - 43807461 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_021424.2 NP_067399.2 2 - 3 494 60
GenBank mRNA AF270977.1 - 2 - 3 426 60
AF297665.1 - 2 - 3 441 60
AK147479.1 - - 1086 60
BC060694.1 - 2 - 3 494 60

View Full Product Details