Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Elavl3
- Entrez Gene ID
-
15571
- Gene Name
- ELAV (embryonic lethal, abnormal vision, Drosophila)-like 3 (Hu antigen C)
- Gene Aliases
-
2600009P04Rik, Huc, PLE21, mHuC
- Chromosome Location
-
Chr.9: 22015005 - 22052023 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK090281.1 |
- |
1 - 2 |
428 |
140 |
View Full Product Details