Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Fat3
- Entrez Gene ID
-
270120
- Gene Name
- FAT tumor suppressor homolog 3 (Drosophila)
- Gene Aliases
-
9430076A06Rik, D430038H04Rik, Gm1132, Gm510, MFAT3F
- Chromosome Location
-
Chr.9: 15910189 - 16502879 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_001080814.1 |
NP_001074283.1 |
15 - 16 |
10213 |
121 |
View Full Product Details