The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_019797.4 | NP_062771.2 | 15 - 16 | 2644 | 83 |
GenBank mRNA | AF197574.1 | - | 12 - 13 | 1703 | 83 |
AF539614.1 | - | 15 - 16 | 2527 | 83 | |
AK146298.1 | - | 12 - 13 | 1711 | 83 | |
BC021316.1 | - | 12 - 13 | 1716 | 83 |