Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rbpms2
Entrez Gene ID
71973
Gene Name
RNA binding protein with multiple splicing 2
Gene Aliases
2400008B06Rik, AI316523
Chromosome Location
Chr.9: 65629336 - 65660536 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_028030.3 NP_082306.2 1 - 2 323 107
GenBank mRNA AK010301.1 - 2 - 3 275 107
AK144659.1 - 1 - 2 348 107
AK156633.1 - 1 - 2 323 107
BC021788.1 - 1 - 2 321 107

View Full Product Details