Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Hpn
Entrez Gene ID
15451
Gene Name
hepsin
Gene Aliases
Hlb320
Chromosome Location
Chr.7: 31098725 - 31115326 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001110252.2 NP_001103722.1 2 - 3 264 87
GenBank mRNA AK002694.1 - 2 - 3 222 87
BC138809.1 - 2 - 3 108 87
BC145413.1 - 2 - 3 108 87

View Full Product Details