Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Zfp27
Entrez Gene ID
22689
Gene Name
zinc finger protein 27
Gene Aliases
Zfp-27, mkr-4, mszf76
Chromosome Location
Chr.7: 29893337 - 29906567 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001037707.1 NP_001032796.1 3 - 4 316 105
NM_001285797.1 NP_001272726.1 3 - 4 588 105
NM_001285798.1 NP_001272727.1 3 - 4 542 105
NM_011754.2 NP_035884.2 3 - 4 311 105
GenBank mRNA AK028500.1 - 3 - 4 543 105
AK135293.1 - 3 - 4 592 105
AK147732.1 - 3 - 4 306 105
BB611831.1 - 3 - 4 296 105
BC059037.1 - 1 - 2 255 105
BY740816.1 - 3 - 4 306 105

View Full Product Details