Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Celf5
Entrez Gene ID
319586
Gene Name
CUGBP, Elav-like family member 5
Gene Aliases
4930565A21Rik, Brunol5
Chromosome Location
Chr.10: 81459227 - 81482725 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_176954.3 NP_795928.2 11 - 12 1278 59
GenBank mRNA AK029796.1 - 11 - 12 2452 59
AK132321.1 - 11 - 12 1244 59
AK158669.1 - 10 - 11 1055 59
BC079845.1 - 11 - 12 1249 59

View Full Product Details