Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Ift57
- Entrez Gene ID
-
73916
- Gene Name
- intraflagellar transport 57
- Gene Aliases
-
4833420A15Rik, Esrrbl1, Hippi, MHS4R2
- Chromosome Location
-
Chr.16: 49699294 - 49765126 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK079748.1 |
- |
- |
781 |
95 |
View Full Product Details