Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
4930545H06Rik
Entrez Gene ID
75148
Gene Name
RIKEN cDNA 4930545H06 gene
Gene Aliases
-
Chromosome Location
Chr.10: 68319065 - 68321142 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_045357.1 1 - 2 86 66
GenBank mRNA AK016046.1 - 1 - 2 87 66


Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BY715634.1 - 1 - 2 83 Chr. 10 - 68319079 - 68321139 [-] on Build GRCm38 66


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream AT rich interactive domain 5B (MRF1-like) Arid5b 4930580B11, 5430435G07Rik, AI467247, Desrt, Mrf2, Mrf2alpha, Mrf2beta -41.5kb Sense Chr. - 68092523 - 68278726 on Build GRCm38
Downstream RIKEN cDNA 1700040L02 gene 1700040L02Rik 109.8kb Sense Chr. - 68430956 - 68541919 on Build GRCm38

View Full Product Details