Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Mei4
Entrez Gene ID
75033
Gene Name
meiosis-specific, MEI4 homolog (S. cerevisiae)
Gene Aliases
4930486G11Rik
Chromosome Location
Chr.9: 81863647 - 82206007 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_175213.3 NP_780422.1 4 - 5 1094 105
GenBank mRNA AK043929.1 - 4 - 5 1095 105
AK166209.1 - 3 - 4 884 105
BC096613.1 - 4 - 5 1136 105

View Full Product Details