The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001205312.1 | NP_001192241.1 | 4 - 5 | 586 | 63 |
NM_018793.2 | NP_061263.2 | 4 - 5 | 586 | 63 | |
GenBank mRNA | AF173032.1 | - | 4 - 5 | 521 | 63 |
AK154438.1 | - | 4 - 5 | 544 | 63 | |
AK155923.1 | - | 4 - 5 | 499 | 63 | |
AK159303.1 | - | 4 - 5 | 555 | 63 | |
AK165483.1 | - | 4 - 5 | 555 | 63 | |
AK167726.1 | - | 4 - 5 | 565 | 63 | |
AK170107.1 | - | 4 - 5 | 525 | 63 | |
AK170940.1 | - | 4 - 5 | 525 | 63 | |
AK171609.1 | - | 4 - 5 | 499 | 63 | |
BC053021.1 | - | 3 - 4 | 491 | 63 | |
BC094240.1 | - | 3 - 4 | 482 | 63 |