Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Icam1
Entrez Gene ID
15894
Gene Name
intercellular adhesion molecule 1
Gene Aliases
CD54, Icam-1, Ly-47, MALA-2
Chromosome Location
Chr.9: 21015960 - 21028797 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_010493.2 NP_034623.1 5 - 6 1267 94
GenBank mRNA AK149748.1 - 5 - 6 1243 94
AK149781.1 - 5 - 6 1242 94
AK149945.1 - 5 - 6 1242 94
AK150003.1 - 5 - 6 1243 94
AK150049.1 - 5 - 6 1242 94
AK150057.1 - 5 - 6 1243 94
AK151227.1 - 5 - 6 1243 94
AK151681.1 - 5 - 6 1243 94
AK152155.1 - 5 - 6 1243 94
AK152527.1 - 5 - 6 1243 94
AK152530.1 - 5 - 6 1243 94
AK152556.1 - 5 - 6 1243 94
AK156417.1 - 5 - 6 1241 94
AK168275.1 - 5 - 6 1243 94
AK171502.1 - 5 - 6 1243 94
AK171520.1 - 4 - 5 980 94
AK172321.1 - 5 - 6 1243 94
CT010246.1 - 5 - 6 1192 94
M31585.1 - 5 - 6 1250 94
X16624.1 - 5 - 6 1221 94
X52264.1 - 5 - 6 1214 94

View Full Product Details