The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001205157.1 | NP_001192086.1 | 1 - 2 | 101 | 71 |
GenBank mRNA | AK033705.1 | - | 1 - 2 | 50 | 71 |
AK133631.1 | - | 1 - 2 | 61 | 71 |