The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_019798.5 | NP_062772.3 | 1 - 2 | 471 | 71 |
GenBank mRNA | AB154250.1 | - | 1 - 2 | 63 | 71 |
AF208021.2 | - | 1 - 2 | 143 | 71 | |
AJ297396.1 | - | 1 - 2 | 143 | 71 |