Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Fxyd7
Entrez Gene ID
57780
Gene Name
FXYD domain-containing ion transport regulator 7
Gene Aliases
1110035I01Rik
Chromosome Location
Chr.7: 31042515 - 31051454 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_022007.1 NP_071290.1 4 - 5 249 64
GenBank mRNA AI892161.1 - 4 - 5 249 64
AK004113.1 - 4 - 5 258 64
BC048737.1 - 4 - 5 262 64

View Full Product Details