Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Mag
Entrez Gene ID
17136
Gene Name
myelin-associated glycoprotein
Gene Aliases
Gma, siglec-4a
Chromosome Location
Chr.7: 30899175 - 30914922 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_010758.2 NP_034888.1 5 - 6 855 76
GenBank mRNA BC101944.1 - 5 - 6 828 76
BC103523.1 - 6 - 7 702 76
M31811.1 - 5 - 6 855 76

View Full Product Details