Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pgr
Entrez Gene ID
18667
Gene Name
progesterone receptor
Gene Aliases
9930019P03Rik, BB114106, ENSMUSG00000074510, NR3C3, PR, PR-A, PR-B
Chromosome Location
Chr.9: 8894736 - 8968611 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_008829.2 NP_032855.2 6 - 7 3108 118
GenBank mRNA AK054117.1 - 6 - 7 1789 118
BC145807.1 - 6 - 7 2513 118
M68915.1 - 6 - 7 2471 118

View Full Product Details