The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001282086.1 | NP_001269015.1 | 1 - 2 | 459 | 65 |
NM_001282087.1 | NP_001269016.1 | 1 - 2 | 459 | 65 | |
NM_013838.2 | NP_038866.2 | 1 - 2 | 459 | 65 | |
GenBank mRNA | AK133864.1 | - | 1 - 2 | 359 | 65 |
BC067391.1 | - | 1 - 2 | 405 | 65 | |
BC068310.1 | - | 1 - 2 | 405 | 65 | |
BC141131.1 | - | 1 - 2 | 330 | 65 | |
U49069.1 | - | 1 - 2 | 459 | 65 |