The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_010421.4 | NP_034551.2 | 11 - 12 | 1388 | 77 |
GenBank mRNA | AK075895.1 | - | 11 - 12 | 1364 | 77 |
AK075911.1 | - | 11 - 12 | 1389 | 77 | |
AK080777.1 | - | 11 - 12 | 1361 | 77 | |
AK144168.1 | - | 11 - 12 | 1516 | 77 | |
AK150444.1 | - | 11 - 12 | 1359 | 77 | |
AK151960.1 | - | 11 - 12 | 1363 | 77 | |
AK159091.1 | - | 12 - 13 | 1359 | 77 | |
AK159814.1 | - | 11 - 12 | 1374 | 77 | |
AK159890.1 | - | 11 - 12 | 1361 | 77 | |
AK160112.1 | - | 11 - 12 | 1348 | 77 | |
AK167013.1 | - | 11 - 12 | 1374 | 77 | |
AK168186.1 | - | 11 - 12 | 1388 | 77 | |
AK170903.1 | - | 11 - 12 | 1375 | 77 | |
AK211327.1 | - | 1 - 2 | 98 | 77 | |
BC010755.1 | - | 11 - 12 | 1357 | 77 |