The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_019482.2 | NP_062355.2 | 4 - 5 | 1645 | 72 |
GenBank mRNA | AF207817.1 | - | 4 - 5 | 1243 | 72 |
AK014033.1 | - | 4 - 5 | 1628 | 72 | |
AK053871.1 | - | 4 - 5 | 1630 | 72 | |
AK089764.1 | - | 4 - 5 | 1591 | 72 | |
AK156194.1 | - | 4 - 5 | 1630 | 72 | |
AK192796.1 | - | 1 - 2 | 57 | 72 | |
AK202295.1 | - | - | 151 | 72 | |
BC020413.1 | - | 4 - 5 | 1576 | 72 | |
BC049074.1 | - | 4 - 5 | 1645 | 72 | |
EU446265.1 | - | 3 - 4 | 1424 | 72 |