Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Vstm5
Entrez Gene ID
69137
Gene Name
V-set and transmembrane domain containing 5
Gene Aliases
2200002K05Rik
Chromosome Location
Chr.9: 15238919 - 15259421 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_026955.2 NP_081231.1 2 - 3 634 98
GenBank mRNA AK008621.1 - 2 - 3 635 98
AK034938.1 - 2 - 3 635 98
AK161045.1 - 2 - 3 427 98
BC055786.1 - 2 - 3 596 98

View Full Product Details