Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Cyp2t4
Entrez Gene ID
384724
Gene Name
cytochrome P450, family 2, subfamily t, polypeptide 4
Gene Aliases
Gm1448, Gm1450
Chromosome Location
Chr.7: 27153714 - 27158973 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001100184.1 NP_001093654.1 3 - 4 370 57

View Full Product Details