Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Hspb6
Entrez Gene ID
243912
Gene Name
heat shock protein, alpha-crystallin-related, B6
Gene Aliases
AA387366, Gm479, Hsp20
Chromosome Location
Chr.7: 30552634 - 30555439 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001012401.2 NP_001012401.1 2 - 3 355 98
GenBank mRNA AK144390.1 - 2 - 3 1010 98
BC051228.1 - 2 - 3 308 98
BC089621.1 - 2 - 3 356 98

View Full Product Details