Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Smg9
Entrez Gene ID
71997
Gene Name
smg-9 homolog, nonsense mediated mRNA decay factor (C. elegans)
Gene Aliases
1500002O20Rik, AI429693, N28092
Chromosome Location
Chr.7: 24398910 - 24422777 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_028047.2 NP_082323.1 5 - 6 862 94
GenBank mRNA AK005126.1 - 5 - 6 862 94
AK039368.1 - 5 - 6 861 94
AK141576.1 - 5 - 6 853 94
AK161454.1 - 5 - 6 872 94
AK169651.1 - 5 - 6 786 94
BC034211.1 - 5 - 6 777 94

View Full Product Details