The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001033324.2 | NP_001028496.1 | 2 - 3 | 1514 | 67 |
GenBank mRNA | AK040211.1 | - | 2 - 3 | 1507 | 67 |
AK142947.1 | - | 2 - 3 | 1515 | 67 | |
BC132441.1 | - | 1 - 2 | 1202 | 67 | |
BC132443.1 | - | 1 - 2 | 1202 | 67 | |
BC138775.1 | - | 1 - 2 | 1325 | 67 | |
Z47205.1 | - | 2 - 3 | 1555 | 67 |