The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_139001.2 | NP_620570.2 | 9 - 10 | 5239 | 74 |
GenBank mRNA | AF352400.1 | - | 9 - 10 | 5183 | 74 |
AK028844.1 | - | 1 - 2 | 457 | 74 | |
AK140986.1 | - | 4 - 5 | 647 | 74 |