Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Alg9
Entrez Gene ID
102580
Gene Name
asparagine-linked glycosylation 9 (alpha 1,2 mannosyltransferase)
Gene Aliases
8230402H15Rik, AI747665, B430313H07Rik, Dibd1
Chromosome Location
Chr.9: 50774881 - 50843639 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_133981.2 NP_598742.1 5 - 6 727 64
NR_045601.1 4 - 5 656 64
GenBank mRNA AK050335.1 - 5 - 6 648 64
AK054293.1 - 5 - 6 633 64
AK086674.1 - 5 - 6 667 64
AK149592.1 - 4 - 5 578 64
AK212096.1 - 3 - 4 234 64
AK214709.1 - 4 - 5 336 64
BC021791.1 - 5 - 6 622 64

View Full Product Details