The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_172896.2 | NP_766484.1 | 5 - 6 | 2323 | 110 |
GenBank mRNA | AK054378.1 | - | 5 - 6 | 2214 | 110 |
AK087843.1 | - | 5 - 6 | 2324 | 110 | |
AK135745.1 | - | 2 - 3 | 1137 | 110 | |
AY355341.1 | - | 5 - 6 | 2324 | 110 | |
AY596200.1 | - | 3 - 4 | 2020 | 110 | |
BC141342.1 | - | 5 - 6 | 2223 | 110 | |
BC141343.1 | - | 5 - 6 | 2223 | 110 |