Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pafah1b3
Entrez Gene ID
18476
Gene Name
platelet-activating factor acetylhydrolase, isoform 1b, subunit 3
Gene Aliases
Pafahg, mus[g]
Chromosome Location
Chr.7: 25295049 - 25297955 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_008776.2 NP_032802.1 5 - 6 540 116
GenBank mRNA AK154547.1 - 5 - 6 536 116
AK167496.1 - 5 - 6 508 116
BC067015.1 - 5 - 6 511 116
U57746.1 - 5 - 6 538 116

View Full Product Details