Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ethe1
Entrez Gene ID
66071
Gene Name
ethylmalonic encephalopathy 1
Gene Aliases
0610025L15Rik, Hsco
Chromosome Location
Chr.7: 24587543 - 24608925 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_023154.3 NP_075643.1 3 - 4 795 83
GenBank mRNA AB049623.1 - 2 - 3 248 83
AK002666.1 - 2 - 3 250 83
BC010592.1 - 2 - 3 253 83
BC083162.1 - 2 - 3 229 83
BC094044.1 - 3 - 4 795 83

View Full Product Details