The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_027875.1 | NP_082151.1 | 6 - 7 | 1657 | 60 |
GenBank mRNA | AK004654.1 | - | 6 - 7 | 1639 | 60 |