The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001081152.1 | NP_001074621.1 | 13 - 14 | 2867 | 74 |
GenBank mRNA | AK033012.1 | - | 13 - 14 | 2847 | 74 |
AK080294.1 | - | 13 - 14 | 2871 | 74 |