Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Adck4
Entrez Gene ID
76889
Gene Name
aarF domain containing kinase 4
Gene Aliases
0610012P18Rik
Chromosome Location
Chr.7: 27228823 - 27257949 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_133770.2 NP_598531.2 14 - 15 1464 95
GenBank mRNA AK002619.1 - 14 - 15 1454 95
AK079366.1 - 4 - 5 508 95
BC013485.1 - 11 - 12 1027 95
BC083324.1 - 12 - 13 1265 95
BC093498.1 - 13 - 14 1401 95

View Full Product Details