Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Odf3l2
Entrez Gene ID
382384
Gene Name
outer dense fiber of sperm tails 3-like 2
Gene Aliases
Gm1157
Chromosome Location
Chr.10: 79639526 - 79655710 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001033473.2 NP_001028645.1 3 - 4 398 86
GenBank mRNA AK158063.1 - 3 - 4 399 86
BC147356.1 - 3 - 4 386 86
BC147357.1 - 3 - 4 386 86
BC172043.1 - 2 - 3 278 86

View Full Product Details