Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Olfr57
Entrez Gene ID
18357
Gene Name
olfactory receptor 57
Gene Aliases
IF12, MOR139-3
Chromosome Location
Chr.10: 79034723 - 79035802 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_147041.2 NP_667252.1 1 - 1 987 94
GenBank mRNA BC141307.1 - 1 - 1 991 94

View Full Product Details