The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001161541.1 | NP_001155013.1 | 1 - 2 | 97 | 84 |
GenBank mRNA | BB611833.1 | - | 1 - 2 | 98 | 84 |